Learn More
5,5,5-Trifluoro-DL-leucine, 94%, Thermo Scientific™
Product Code. 11373426
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373426 100 mg 100mg
11363426 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11373426 Supplier Thermo Scientific Alfa Aesar Supplier No. L12890.MC
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5,5,5-Trifluoro-DL-leucine is used as an intermediate in chemical research.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at -80°C
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2792-72-5
Molecular Formula C6H10F3NO2
Molecular Weight (g/mol) 185.146
MDL Number MFCD00039528
InChI Key XFGVJLGVINCWDP-UHFFFAOYSA-N
Synonym 5,5,5-trifluoro-dl-leucine, 5,5,5-trifluoroleucine, 5',5',5'-trifluoroleucine, dl-5',5',5'-trifluoroleucine, leucine,5,5,5-trifluoro, leucine, 5,5,5-trifluoro, 2-azanyl-5,5,5-tris fluoranyl-4-methyl-pentanoic acid, acmc-1cocp, norvaline, 5,5,5-trifluoro-4-methyl-, dl, dl-leucine, 5,5,5-trifluoro
PubChem CID 259832
ChEBI CHEBI:78014
IUPAC Name 2-amino-5,5,5-trifluoro-4-methylpentanoic acid
SMILES CC(CC(C(=O)O)N)C(F)(F)F
Specifications
Melting Point 273°C to 275°C
Beilstein 4664821
Quantity 100 mg
Formula Weight 185.15
Percent Purity 94%
Chemical Name or Material 5,5,5-Trifluoro-DL-leucine