missing translation for 'onlineSavingsMsg'
Learn More

5-[2-(Trifluoromethyl)phenyl]-1,2-oxazole-4-carboxylic Acid, TRC

Artikelnummer. 30456428
Change view
Click to view available options
Menge:
10 mg
50 mg
Packungsgröße:
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30456428 50 mg 50 Milligramm
30456455 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30456428 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCT79212550MG

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1003561-80-5
Summenformel C11H6NO3F3
Molekulargewicht (g/mol) 257.17
IUPAC-Name 5-[2-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxylic acid
SMILES OC(=O)c1cnoc1c2ccccc2C(F)(F)F

Spezifikation

Chemischer Name oder Material 5-[2-(Trifluoromethyl)phenyl]-1,2-oxazole-4-carboxylic Acid
Menge 50 mg
InChI-Formel InChI=1S/C11H6F3NO3/c12-11(13,14)8-4-2-1-3-6(8)9-7(10(16)17)5-15-18-9/h1-5H,(H,16,17)
Formelmasse 257.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.