Learn More
Agilent BioTek 406 FX Product Qualification Package
406 FX Product Qualification Package for use with the 406 FX Washer Dispenser.
Marke:  Agilent BioTek 2120530
Additional Details : Gewicht : 2.26796kg
Artikelnummer. 17824657
  • 1485.00 CHF / 1 Stück
Verfügbar ab: 28-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Includes thorough procedures to allow a user to perform Installation Qualification, Operational Qualification and Performance Qualification
  • Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance
  • Product Qualification Packages contain only documentation and procedures
  • Many of these procedures require the use of QC Test Solutions or other accessories, and these are ordered separately
Spezifikation
Spezifikation
Service Kit
Agilent BioTek 406 FX Washer Dispenser