Learn More
4-(Trifluoromethylthio)phenol, 98%, Thermo Scientific™
Product Code. 11421587
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421587 5 g 5g
11411587 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11421587 Supplier Thermo Scientific Alfa Aesar Supplier No. B24996.06
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 461-84-7
Molecular Formula C7H5F3OS
Molecular Weight (g/mol) 194.171
MDL Number MFCD00036035
InChI Key YYCPTWHVKSATQK-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio phenol, 4-trifluoromethyl thio phenol, 4-trifluoromethyl sulfanyl phenol, 4-trifluoromethylsulfanyl-phenol, 4-trifluoromethylsulfanyl phenol, 4-trifluoromethylthio hydroxybenzene, p-trifluoromethylthio phenol, 4-trifluoromethyl sulphanyl phenol, phenol, 4-trifluoromethyl thio
PubChem CID 3815158
IUPAC Name 4-(trifluoromethylsulfanyl)phenol
SMILES C1=CC(=CC=C1O)SC(F)(F)F
Specifications
Melting Point 57°C to 60°C
Boiling Point 77°C to 78°C (7 mmHg)
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 194.18
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethylthio)phenol