Learn More
4-(Trifluormethyl)benzhydrazid, TRC
Specifications
Specifications
| Chemischer Name oder Material | 4-(Trifluoromethyl)benzhydrazide |
| CAS | 339-59-3 |
| Summenformel | C9 H6 O2 S2 |
| Menge | 1 g |
| Synonym | 4-(Trifluoromethyl)benzohydrazide, 4-(Trifluoromethyl)benzoic Hydrazide, 4-Trifluoromethylbenzhydrazide, 4-Trifluoromethylbenzoic Acid Hydrazide, p-(Trifluoromethyl)benzoylhydrazine, p-Trifluoromethylbenzoic Acid Hydrazide, p-Trifluoromethylbenzoyl Hydrazide |
| InChI-Formel | InChI=1S/C9H6O2S2/c10-9(11)8-4-3-7(13-8)6-2-1-5-12-6/h1-5H,(H,10,11) |
| SMILES | OC(=O)c1ccc(s1)c2cccs2 |
| IUPAC-Name | 5-Thiophen-2-ylthiophen-2-Carbonsäure |
| Molekulargewicht (g/mol) | 210.273 |
| Formelmasse | 209.981 |