Learn More
4-(Methylthio)imidazol, 97 %, Thermo Scientific Chemicals
Catalog Number p-7041681
Click to view available options
Menge:
250 mg
1 g
83553-60-0
C4H6N2S
114.166
MFCD03844702
NLIJIXLRIYPOHM-UHFFFAOYSA-N
4-methylthio-1h-imidazole, 4-methylthio imidazole, 5-methylthio-1h-imidazole, 4-methylsulfanyl-1h-imidazole, pubchem14136, 4-methylthio ;-1h-imidazole, 5-methylsulfanyl-1h-imidazole, 1h-imidazole,4-methylthio-9ci, 1h-imidazole, 4-methylthio
13084405
5-Methylsulfanyl-1H-Imidazol
CSC1=CN=CN1
83553-60-0
C4H6N2S
114.166
MFCD03844702
NLIJIXLRIYPOHM-UHFFFAOYSA-N
4-methylthio-1h-imidazole, 4-methylthio imidazole, 5-methylthio-1h-imidazole, 4-methylsulfanyl-1h-imidazole, pubchem14136, 4-methylthio ;-1h-imidazole, 5-methylsulfanyl-1h-imidazole, 1h-imidazole,4-methylthio-9ci, 1h-imidazole, 4-methylthio
13084405
5-Methylsulfanyl-1H-Imidazol
CSC1=CN=CN1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Methylthio)imidazole
Menge 250 mg
Formelmasse 114.17
Reinheit (%) 97%