missing translation for 'onlineSavingsMsg'
Learn More

4-(Furfurylaminocarbonyl)-benzolboronsäurepinakolester, 97 %, Thermo Scientific™

Catalog Number p-7030507
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-59-9
C18H22BNO4
327.19
MFCD09266168
ZNYVQYJVECCENX-UHFFFAOYSA-N
4-furfurylaminocarbonyl phenylboronic acid pinacol ester, n-furan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, n-furan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb018, 4-furfurylaminocarbonyl benzeneboronic acid pinacol ester, 4-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-ylmethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739034
N-(2-ylmethyl)-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Benzamid
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NCC1=CC=CO1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-59-9
C18H22BNO4
327.19
MFCD09266168
ZNYVQYJVECCENX-UHFFFAOYSA-N
4-furfurylaminocarbonyl phenylboronic acid pinacol ester, n-furan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, n-furan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb018, 4-furfurylaminocarbonyl benzeneboronic acid pinacol ester, 4-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-ylmethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739034
N-(2-ylmethyl)-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Benzamid
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NCC1=CC=CO1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
Schmelzpunkt 113°C to 114°C
Menge 1 g
Formelmasse 327.18
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'