missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenylisothiocyanat, 97 %, Thermo Scientific Chemicals

Catalog Number p-7027530
Click to view available options
Menge:
2 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
18856-63-8
C9H9NS
163.24
MFCD00041101
XXLWCGRHJFEMQH-UHFFFAOYSA-N
4-ethylphenyl isothiocyanate, 1-ethyl-4-isothiocyanato-benzene, benzene,1-ethyl-4-isothiocyanato, 4-ethylbenzenisothiocyanate, acmc-20amit, 2-p-isothio-cyanatophenyl ethane, #
140438
1-Ethyl-4-isothiocyanatobenzol
CCC1=CC=C(C=C1)N=C=S
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
18856-63-8
C9H9NS
163.24
MFCD00041101
XXLWCGRHJFEMQH-UHFFFAOYSA-N
4-ethylphenyl isothiocyanate, 1-ethyl-4-isothiocyanato-benzene, benzene,1-ethyl-4-isothiocyanato, 4-ethylbenzenisothiocyanate, acmc-20amit, 2-p-isothio-cyanatophenyl ethane, #
140438
1-Ethyl-4-isothiocyanatobenzol
CCC1=CC=C(C=C1)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Ethylphenyl isothiocyanate
Siedepunkt 245°C
Flammpunkt >110°C (230°F)
Geruch Pungent
Menge 2 g
UN-Nummer UN2922
Beilstein 2802384
Formelmasse 163.24
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'