Learn More
4-Biphenylylcarboxaldehyd, TRC
Specifications
Specifications
| Chemischer Name oder Material | 4-Biphenylylcarboxaldehyde |
| CAS | 3218-36-8 |
| Summenformel | C13H10O |
| Menge | 10 g |
| Synonym | 4-Biphenylcarboxaldehyde, p-Phenyl-benzaldehyde, 4-Biphenylaldehyde, 4-Biphenylylcarboxaldehyde, 4-Formyl-1,1'-biphenyl, 4-Formylbiphenyl, 4-Phenylbenzaldehyde, Biphenyl-4-carbaldehyde, NSC 46066, p-Biphenylaldehyde, p-Biphenylcarboxaldehyde, p-Biphenylylaldehyde, p-Phenylbenzaldehyde |
| InChI-Formel | InChI=1S/C13H10O/c14-10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-10H |
| SMILES | O=Cc1ccc(cc1)c2ccccc2 |
| IUPAC-Name | 4-Phenylbenzaldehyd |
| Molekulargewicht (g/mol) | 182.22 |
| Formelmasse | 182.0732 |