missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl, 98 %, Thermo Scientific Chemicals

Catalog Number 11408446
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
787-69-9
C16H14O2
238.286
MFCD00017248
YSTSBXDVNKYPTR-UHFFFAOYSA-N
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
301558
1-[4-(4-Acetylphenyl)phenyl]ethanon
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
787-69-9
C16H14O2
238.286
MFCD00017248
YSTSBXDVNKYPTR-UHFFFAOYSA-N
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
301558
1-[4-(4-Acetylphenyl)phenyl]ethanon
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4,4'-Diacetylbiphenyl
Schmelzpunkt 190°C to 193°C
Menge 5 g
Formelmasse 238.29
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'