Learn More
4-1BB/TNFRSF9/CD137 Antibody (2356B) [Janelia Fluor™ 549], Novus Biologicals™
Rabbit Monoclonal Antibody
Marke:  Novus Biologicals FAB8381I
Artikelnummer. 18579447
  • 744.00 CHF / 0.10 Milliliter
Definieren Sie Ihre Immunologie-Forschung nach Ihren Bedingungen
Verwenden Sie den Aktionscode 24090
Alle Werbeaktionen anzeigen
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
4-1BB/TNFRSF9/CD137
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry, CyTOF
2356B
Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry, CyTOF-ready
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Chinese hamster ovary cell line CHO-derived recombinant human 4-1BB/TNFRSF9/CD137, Leu24-His183, Accession # Q07011
0.1 mL
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
3604
Store at 4°C in the dark.
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen