Learn More
3M™ PU-Leichtschlauch mit Stahlspirale, kurz Länge: 73,5 cm
Short, lightweight breathing tube, easy to wipe down. 3M™ Versaflo™ BT-Series Compatible Breathing Tubes are compatible with all 3M™ Versaflo™ Headtops and Air Delivery Units.
Marke: 3M™ 7000000158
Additional Details : Gewicht : 0.80000kg
Beschreibung
- Short, lightweight breathing tube that is easy to wipe down
- Quick release swivel enables one-handed connection and release.
- Disposable breathing tube cover is available to help cleaning and decontamination operations