Learn More
3-Piperidinopropiophenone Hydrochloride, TRC
Artikelnummer. 17903595
Change view
Click to view available options
Menge:
5 g
Packungsgröße:
5 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17903595 5 g 5 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17903595 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCP4820105G
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 886-06-6
Summenformel C14 H19 N O . Cl H
Molekulargewicht (g/mol) 253.77
Synonym 1-Phenyl-3-(piperidin-1-yl)propan-1-one Hydrochloride, Trihexyphenidyl Hydrochloride Imp. A (EP) as Hydrochloride, Piperidyl Propiophenone Hydrochloride, Pridinol Hydrochloride Imp. A as Hydrochloride, 1-Propanone, 1-phenyl-3-(1-piperidinyl)-, hydrochloride (1:1), 1-Propanone, 1-phenyl-3-(1-piperidinyl)-, hydrochloride (9CI), Propiophenone, 3-piperidino-, hydrochloride (6CI,7CI,8CI), 1-(2-Benzoylethyl)piperidinium chloride, 1-(3-Oxo-3-phenylpropyl)piperidin-1-ium chloride, 1-(3-Oxo-3-phenylpropyl)piperidinium chloride, 1-Phenyl-3-(piperidin-1-yl)propan-1-one hydrochloride, 3-(1-Piperidinyl)propiophenone hydrochloride, 3-Piperidinopropiophenone hydrochloride, N-(2-Benzoylethyl)piperidine hydrochloride, S 220, β-Piperidinopropiophenone hydrochloride
IUPAC-Name 1-phenyl-3-piperidin-1-ylpropan-1-one;hydrochloride
SMILES Cl.O=C(CCN1CCCCC1)c2ccccc2
Spezifikation
Chemischer Name oder Material 1-Phenyl-3-(piperidin-1-yl)propan-1-one Hydrochloride
Menge 5 g
InChI-Formel InChI=1S/C14H19NO.ClH/c16-14(13-7-3-1-4-8-13)9-12-15-10-5-2-6-11-15;/h1,3-4,7-8H,2,5-6,9-12H2;1H
Formelmasse 253.1233