Learn More
R&D Systems™ 3'-Phosphoadenosine-5'-phosphosulfate
Artikelnummer. 18782464
Ansicht ändern
Click to view available options
Menge:
500 μg
Packungsgröße:
500 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18782464 500 μg 500 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18782464 Supplier R&D Systems™ Supplier No. ES019
  • ONLINE EXCLUSIVE PRICE
    287.00 CHF
    270.90 CHF / 500 Mikrogramm
    Sparen 16.10 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Substrate for sulfotransferase assay
3'-Phosphoadenosine-5'-phosphosulfate (PAPS) is the universal sulfur donor for sulfotransferases. It is synthesized from ATP through two steps. In the first step, one ATP molecule is converted to adenosine-5'-phosphosulfate (APS) by ATP sulfurylase and pyrophosphate (PPi) is generated as a byproduct. In the second step, the APS is converted to PAPS by APS kinase and ADP is generated as a byproduct. To drive the reaction forward, the pyrophosphate can be further degraded to phosphate (Pi) by inorganic pyrophosphatase to eliminate feedback inhibition, and the ADP can be converted back to ATP using pyruvate kinase.
  • Suitable for kinetic assay of sulfotransferases using the Universal Sulfotransferase Activity Kit (Mfr. No. EA003) or for enzymatic synthesis of sulfonated molecules
  • Purity >90% of the nucleotide based on HPLC: No Li+ or Na+ ions, and free inorganic phosphate <0.5%
TRUSTED_SUSTAINABILITY
Specifications
Menge 500 μg
Produkttyp PAPS, C10H15N5O13P2S
Lagerungsbedingungen Store the unopened product at -70° C for up to six months. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.