Learn More
TERUMO 3-Part Syringes
Click to view available options
Volumen (métrico):
1 mL
2.5 mL
3 mL
5 mL
10 mL
30 mL
Tipo de conexión:
Centric Luer Lock
Centric Luer Slip
Concentric Luer Lock
Eccentric Luer Slip
Eccentric Luer Tip
Luer Slip
Packungsgröße:
100 Stück
25 Stück
Beschreibung
- Every single detail is the result of Terumo's long time experience in this field which is appreciated by users throughout the world
- Siliconisation for optimal plunger mobility
- The integral gasket ensures a fi rm and distinct end-stop
- Clear graduated scale allows accurate dosage
- Transparent barrel for visualisation
- Finger grips to support easy handling
Spezifikation
Spezifikation
Esterilizables en autoclave | Not Autoclavable |
Material | Polypropylene (Barrel), Polypropylene (Plunger) |
Sin látex | Yes |
Tipo de producto | Syringe |
Desechables | Single-use |
Graduaciones | 0.01 mL |
Volumen (métrico) | 1 mL |
Tipo de conexión | Luer Slip |
Graduated | Yes |
Esterilidad | Sterile |
Berichtigung von Produktinhalten
Name des Produkts
TERUMO 3-Part Syringes >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur