Learn More
3-O-Methyl-D-Glucose Kristallines Pulver MP Biomedicals
Artikelnummer. 11462372 Alle ansehen MP Biomedicals Produkte
Change view
Click to view available options
Menge:
5 g
25 g
Packungsgröße:
25 Gramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11462372 5 g -
11482372 25 g 25 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11462372 Lieferant MP Biomedicals Lieferanten-Nr. 0215165105
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Chemische Identifikatoren
CAS 13224-94-7
Summenformel C7H14O6
Molekulargewicht (g/mol) 194.18
MDL-Nummer MFCD00063266
InChI-Schlüssel SCBBSJMAPKXHAH-OVHBTUCOSA-N
Synonym 3-o-methyl-alpha-d-glucopyranose, 3-o-methyl-a-d-glucopyranose, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxytetrahydro-2h-pyran-2,3,5-triol, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxyoxane-2,3,5-triol, 3-o-methyl-, a-d-glucopyranose, 3-o-methyl-alpha-d-glucose, a-d-glucopyranose,3-o-methyl, alpha-d-glucopyranose, 3-o-methyl, 3-o-methyl-d-glucopyranose
PubChem CID 83246
IUPAC-Name (2S,3R,4S,5R,6R)-6-(hydroxymethyl)-4-methoxyoxan-2,3,5-triol
SMILES CO[C@@H]1[C@@H](O)[C@@H](O)O[C@H](CO)[C@H]1O
Spezifikation
Chemischer Name oder Material 3-O-methyl-D-glucose
Farbe Weiß
Menge 5 g
Formelmasse 194.2
Physikalische Form Pulver