missing translation for 'onlineSavingsMsg'
Learn More

3-Fluor-2-methylbenzonitril, 97 %

Catalog Number 11431370
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
185147-06-2
C8H6FN
135.14
MFCD03788541
DMQFTNHJWVKVGE-UHFFFAOYSA-N
2-cyano-6-fluorotoluene, 2-methyl-3-fluoro benzonitrile, benzonitrile, 3-fluoro-2-methyl, 6-cyano-2-fluorotoluene, 3-fluoro-2-methylbenzenecarbonitrile, pubchem1556, acmc-209emm, ksc497i2n, 2-methyl-3-fluorobenzonitrile, 3-fluoro-2-methyl-benzonitrile
2779238
3-fluoro-2-methylbenzonitrile
CC1=C(F)C=CC=C1C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
185147-06-2
C8H6FN
135.14
MFCD03788541
DMQFTNHJWVKVGE-UHFFFAOYSA-N
2-cyano-6-fluorotoluene, 2-methyl-3-fluoro benzonitrile, benzonitrile, 3-fluoro-2-methyl, 6-cyano-2-fluorotoluene, 3-fluoro-2-methylbenzenecarbonitrile, pubchem1556, acmc-209emm, ksc497i2n, 2-methyl-3-fluorobenzonitrile, 3-fluoro-2-methyl-benzonitrile
2779238
3-fluoro-2-methylbenzonitrile
CC1=C(F)C=CC=C1C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Fluoro-2-methylbenzonitrile
Brechungsindex 1.509
Menge 1 g
UN-Nummer UN3276
Formelmasse 135.14
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'