missing translation for 'onlineSavingsMsg'
Learn More

3-(Dichloroacetyl)-2,2,5-trimethyloxazolidine, TRC

Artikelnummer. 30180184
Change view
Click to view available options
Menge:
100 mg
Packungsgröße:
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30180184 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30180184 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD666955100MG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 52836-31-4
Summenformel C8 H13 Cl2 N O2
Molekulargewicht (g/mol) 226.1
Synonym Oxazolidine, 3-(dichloroacetyl)-2,2,5-trimethyl- (9CI), 2,2-Dichloro-1-(2,2,5-trimethyl-3-oxazolidinyl)ethanone, 2,2,5-Trimethyl-3-(dichloroacetyl)-1,3-oxazolidine, 2,2,5-Trimethyl-3-(dichloroacetyl)oxazolidine, R 29148
IUPAC-Name 2,2-dichloro-1-(2,2,5-trimethyl-1,3-oxazolidin-3-yl)ethanone
SMILES CC1CN(C(=O)C(Cl)Cl)C(C)(C)O1

Specifications

Chemischer Name oder Material 3-(dichloroacetyl)-2,2,5-trimethyloxazolidine
Menge 100 mg
InChI-Formel InChI=1S/C8H13Cl2NO2/c1-5-4-11(7(12)6(9)10)8(2,3)13-5/h5-6H,4H2,1-3H3
Formelmasse 225.0323
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.