Learn More
3-Des[4-(2-Piperidinyl)ethoxy]benzoyl-7-[4-(2-Piperidinyl)ethoxy]benzoyl Raloxifene (>90%), TRC
Artikelnummer. 30367449
Change view
Click to view available options
Menge:
1 mg
10 mg
25 mg
Packungsgröße:
1 Milligramm
10 Milligramm
25 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30367449 1 mg 1 Milligramm
30351933 25 mg 25 Milligramm
30349921 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367449 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD2970501MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 1391054-73-1
Summenformel C28 H27 N O4 S
Molekulargewicht (g/mol) 473.58
Synonym Methanone, [6-hydroxy-2-(4-hydroxyphenyl)benzo[b]thien-7-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]-, [6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothien-7-yl][4-(2-piperidin-1-yl)ethoxy]-phenyl]methanone, Raloxifene Hydrochloride Imp. B (EP)
IUPAC-Name [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-7-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone
SMILES Oc1ccc(cc1)c2cc3ccc(O)c(C(=O)c4ccc(OCCN5CCCCC5)cc4)c3s2
Spezifikation
Chemischer Name oder Material 3-Des[4-(2-Piperidinyl)ethoxy]benzoyl-7-[4-(2-Piperidinyl)ethoxy]benzoyl Raloxifene
Menge 1 mg
InChI-Formel InChI=1S/C28H27NO4S/c30-22-9-4-19(5-10-22)25-18-21-8-13-24(31)26(28(21)34-25)27(32)20-6-11-23(12-7-20)33-17-16-29-14-2-1-3-15-29/h4-13,18,30-31H,1-3,14-17H2
Formelmasse 473.1661