Learn More
3-Chlorobisphenol A, TRC
Artikelnummer. 30005791
Change view
Click to view available options
Menge:
5 mg
25 mg
Packungsgröße:
25 Milligramm
5 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30005791 5 mg 5 Milligramm
17954215 25 mg 25 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005791 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCC3645805MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 74192-35-1
Summenformel C15 H15 Cl O2
Molekulargewicht (g/mol) 262.73
Synonym 2-Chloro-4-[1-(4-hydroxyphenyl)-1-methylethyl]phenol, 2-Chloro-4,4'-isopropylidenediphenol, Chlorobisphenol A
IUPAC-Name 2-chloro-4-[1-(4-hydroxyphenyl)-1-methyl-ethyl]phenol
SMILES CC(C)(c1ccc(O)cc1)c2ccc(O)c(Cl)c2
Spezifikation
Chemischer Name oder Material 3-Chlorobisphenol A
Hinweise zur Reinheitsqualität HPLC
Menge 5 mg
InChI-Formel InChI=1S/C15H15ClO2/c1-15(2,10-3-6-12(17)7-4-10)11-5-8-14(18)13(16)9-11/h3-9,17-18H,1-2H3
Formelmasse 262.0761
Reinheit (%) >95%