missing translation for 'onlineSavingsMsg'
Learn More

3,6-Thioxanthenediamine-10,10-dioxide, 97%

Product Code. 10519102 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10519102 25 g Glass bottle 25g
10683732 1 g Glass bottle 1g
10710302 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10519102 Supplier Thermo Scientific Acros Supplier No. 420910250
 Restricted Item
Lager vorrätig
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10215-25-5 | C13H12N2O2S | 260.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10215-25-5
Molecular Formula C13H12N2O2S
Molecular Weight (g/mol) 260.32
MDL Number MFCD00041841
InChI Key UPVRZVIJGVFROW-UHFFFAOYSA-N
PubChem CID 82456
IUPAC Name 10,10-dioxo-9H-thioxanthene-3,6-diamine
SMILES C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 213°C to 215°C
Color Beige to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Synonym 9h-thioxanthene-3,6-diamine 10,10-dioxide, 9h-thioxanthene-3,6-diamine, 10,10-dioxide, 3,6-diamino-9h-thioxanthene 10,10-dioxide, 3,6-thioxanthenediamine-10,10-dioxide, 3,7-thiaxanthenediamine-5,5-dioxide, 9h-thioxanthene-3,7-diamine, 5,5-dioxide, 9h-thioxanthene-3,6-diamine,10,10-dioxide, 3,7-diamino-10h-dibenzo b,e thiin-5,5-dione, 3,6-diamino-9h-10??-thioxanthene-10,10-dione, 3,6-diamino-9h-10
Formula Weight 260.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3, 6-Thioxanthenediamine-10, 10-dioxide, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.