missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrosalicylsäure, TRC

Product Code. 30006888
Change view
Click to view available options
Menge:
5 g
25 g
Unit Size:
25 Gramm
5 Gramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30006888 5 g 5 Gramm
30010545 25 g 25 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006888 Supplier Toronto Research Chemicals Supplier No. TRCD4803005G
 Eingeschränkter Artikel
Verfügbar ab: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-99-4
Summenformel C7 H4 N2 O7
Molekulargewicht (g/mol) 228.12
Synonym 3,5-Dinitrosalicylate, 3,5-Dinitro-2-hydroxybenzoic acid
IUPAC-Name 2-Hydroxy-3,5-Dinitrobenzoesäure
SMILES OC(=O)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Chemischer Name oder Material 3,5-Dinitrosalicylic Acid
Menge 5 g
InChI-Formel InChI=1S/C7H4N2O7/c10-6-4(7(11)12)1-3(8(13)14)2-5(6)9(15)16/h1-2,10H,(H,11,12)
Formelmasse 228.0019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.