missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylendioxy)-benzylalkohol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7020635
Click to view available options
Menge:
25 g
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
495-76-1
C8H8O3
152.149
MFCD00005836
BHUIUXNAPJIDOG-UHFFFAOYSA-N
piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
10322
1,3-Benzodioxol-5-ylmethanol
C1OC2=C(O1)C=C(C=C2)CO
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
495-76-1
C8H8O3
152.149
MFCD00005836
BHUIUXNAPJIDOG-UHFFFAOYSA-N
piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
10322
1,3-Benzodioxol-5-ylmethanol
C1OC2=C(O1)C=C(C=C2)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,4-(Methylenedioxy)benzyl alcohol
Schmelzpunkt 50°C to 55°C
Siedepunkt 156°C to 158°C (16 mmHg)
Flammpunkt >110°C (230°F)
Menge 25 g
Beilstein 136113
Formelmasse 152.15
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'