missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminodipropylamine, 99+%, Thermo Scientific™

Artikelnummer. 10235210 Alle ansehen Thermo Scientific Chemicals Produkte
Ansicht ändern
Click to view available options
Menge:
100 g
Verpackung:
Glasflasche
Packungsgröße:
100 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Verpackung unitSize
10235210 100 g Glasflasche 100 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10235210 Lieferant Thermo Scientific Acros Lieferanten-Nr. 122051000
Bulk oder benutzerdefiniertes Format anfordern

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 56-18-8
Summenformel C6H17N3
Molekulargewicht (g/mol) 131.223
MDL-Nummer MFCD00008214
InChI-Schlüssel OTBHHUPVCYLGQO-UHFFFAOYSA-N
Synonym norspermidine, bis 3-aminopropyl amine, dipropylenetriamine, 3,3'-diaminodipropylamine, 3,3'-iminobispropylamine, caldine, 3,3'-iminobis propylamine, 1,7-diamino-4-azaheptane, 4-azaheptane-1,7-diamine, 3,3-diaminodipropylamine
PubChem CID 5942
ChEBI CHEBI:16841
IUPAC-Name N'-(3-aminopropyl)propane-1,3-diamine
SMILES C(CN)CNCCCN

Specifications

Chemischer Name oder Material 3, 3'-Diaminodipropylamine
Namenshinweis 98%
CAS Min. % 99.0
CAS Max. % 100.0
Farbe Straw to Yellow
Dichte 0.9300g/mL
Infrarotspektrum Authentic
Prozentgehaltsbereich 99+%
Verpackung Glasflasche
Strukturformel HN[(CH2)3NH2]2
Brechungsindex 1.4810 to 1.4830 (20°C, 589nm)
Menge 100 g
Formelmasse 131.22
Reinheit (%) ≥99.0%
Physikalische Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.