missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenoxy)-1,2-propandiol, 99+%, Thermo Scientific Chemicals

Catalog Number p-7023365
Click to view available options
Menge:
50 g
250 g
1000 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
93-14-1
C10H14O4
198.22
MFCD00016873
HSRJKNPTNIJEKV-UHFFFAOYNA-N
guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol
3516
3-(2-methoxyphenoxy)propane-1,2-diol
COC1=CC=CC=C1OCC(O)CO
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
93-14-1
C10H14O4
198.22
MFCD00016873
HSRJKNPTNIJEKV-UHFFFAOYNA-N
guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol
3516
3-(2-methoxyphenoxy)propane-1,2-diol
COC1=CC=CC=C1OCC(O)CO

3-(2-Methoxyphenoxy)-1,2-propanediol is involved in centrally acting muscle relaxant with expectorant properties. It is often used as anesthetic during animal surgery. It is used in the treatment of coughing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Löslich in Wasser (50 mg/ml bei 25 °C), Methanol, DMSO (40 mg/ml bei 25 °C), DMF, Ethanol (50 mg/ml), Glycerin und Glykol.

Hinweise
Kühl und trocken in einem gut abgedichteten Behälter lagern. Von Oxidationsmitteln entfernt aufbewahren
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(2-Methoxyphenoxy)-1,2-propanediol
Schmelzpunkt 80°C to 82°C
Siedepunkt 215°C (19 mmHg)
Menge 50 g
Beilstein 2049375
Merck Index 14,4555
Löslichkeitsinformationen Soluble in water (50mg/ml at 25°C),methanol,DMSO (40mg/ml at 25°C),DMF,ethanol (50mg/ml),glycerol and glycol.
Formelmasse 198.22
Reinheit (%) ≥99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'