missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-Imidazol-1-yl)-propansäure, ≥ 97 %, Thermo Scientific™
Click to view available options
Menge:
250 mg
1 g
5 g
Spezifikation
Chemischer Name oder Material | 3-(1H-imidazol-1-yl)propanoic acid |
CAS Min. % | 97.0 |
Prozentgehaltsbereich | 97% |
Menge | 5 g |
Formelmasse | 140.14 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'