Learn More
2-(Trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™
Product Code. 11467597
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467597 25 g 25g
11447597 1 g 1g
11457597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11467597 Supplier Thermo Scientific Alfa Aesar Supplier No. A15667.14
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 346-06-5
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.14
MDL Number MFCD00004621
InChI Key TWQNSHZTQSLJEE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol
PubChem CID 67666
IUPAC Name [2-(trifluoromethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1C(F)(F)F
Specifications
Melting Point 4°C
Density 1.326
Boiling Point 80°C to 83°C (3 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.468
Quantity 25 g
Beilstein 2093714
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzyl alcohol