Learn More
Masterflex™ 2-Stop, PVC-Stopped Tubing with Flared Ends
Phthalate-free tubing with a long pumping life and a wide range of acid and chemical resistance.
Marke: Masterflex™ 97616-10
Additional Details : Gewicht : 0.04536kg
Beschreibung
- USP Class VI tubing
- Phthalate-free
- Long pumping life
- Applications include laboratory, pharmaceutical, peristaltic pumps.
- 61 Shore A
- REACH and RoHS compliant
- Very low gas permeability and can withstand high pressure contents.
- Plasticizer is very tightly bonded into the material, preventing any leaching of component chemicals into the medium being transported, maintaining its sterility.
- Transparent for visual observation of tubing contents.
Spezifikation
3A;ADCF;European Pharmacopoeia;Non-DEHP;FDA;Non-Phthalate;NSF-51;USP;NSF-61 | |
0.19 mm | |
0.381 m | |
I.D. 0.19 mm |
Orange-Red | |
Non-DEHP PVC | |
12/Pk |