Learn More
2-[(Phenylsulfanyl)methyl]benzolcarbonsäure, TRC
Specifications
Specifications
| Chemischer Name oder Material | 2-[(Phenylsulfanyl)methyl]benzenecarboxylic Acid |
| CAS | 1699-03-2 |
| Summenformel | C14H12O2S |
| Menge | 50 mg |
| Synonym | o-Toluic acid, alpha-(phenylthio)- (6CI,7CI,8CI), 2-[(Phenylthio)methyl]benzoic acid, 2-(Phenylthiomethyl)benzoic acid, 2-[(Phenylsulfanyl)methyl]benzoic acid, alpha-(Phenylthio)-o-toluic acid |
| InChI-Formel | InChI=1S/C14H12O2S/c15-14(16)13-9-5-4-6-11(13)10-17-12-7-2-1-3-8-12/h1-9H,10H2,(H,15,16) |
| SMILES | OC(=O)c1ccccc1CSc2ccccc2 |
| IUPAC-Name | 2-(Phenylsulfanylmethyl)benzoesäure |
| Molekulargewicht (g/mol) | 244.31 |
| Formelmasse | 244.0558 |