missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzylbromid, 98 %, Thermo Scientific Chemicals

Catalog Number 11490317
Click to view available options
Menge:
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
89-92-9
C8H9Br
185.064
MFCD00000174
WGVYCXYGPNNUQA-UHFFFAOYSA-N
2-methylbenzyl bromide, 1-bromomethyl-2-methylbenzene, alpha-bromo-o-xylene, o-xylyl bromide, o-methylbenzyl bromide, 2-xylyl bromide, 2-bromomethyl toluene, 2-methylbenzylbromide, benzene, 1-bromomethyl-2-methyl, a-bromo-o-xylene
6992
1-(Brommethyl)-2-Methylbenzol
CC1=CC=CC=C1CBr
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
89-92-9
C8H9Br
185.064
MFCD00000174
WGVYCXYGPNNUQA-UHFFFAOYSA-N
2-methylbenzyl bromide, 1-bromomethyl-2-methylbenzene, alpha-bromo-o-xylene, o-xylyl bromide, o-methylbenzyl bromide, 2-xylyl bromide, 2-bromomethyl toluene, 2-methylbenzylbromide, benzene, 1-bromomethyl-2-methyl, a-bromo-o-xylene
6992
1-(Brommethyl)-2-Methylbenzol
CC1=CC=CC=C1CBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methylbenzyl bromide
Schmelzpunkt 18°C to 20°C
Dichte 1.381
Siedepunkt 216°C to 217°C
Flammpunkt 82°C (179°F)
Brechungsindex 1.575
Menge 25 g
UN-Nummer UN1701
Beilstein 636333
Merck Index 14,10090
Formelmasse 185.07
Reinheit (%) 98%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'