missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxyflavanon, 98 %, Thermo Scientific Chemicals

Catalog Number p-7027958
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
35244-11-2
C15H12O3
MFCD00017702
2r-2-2-hydroxyphenyl-2,3-dihydrochromen-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, 2r-2-2-hydroxyphenyl-3,4-dihydro-2h-1-benzopyran-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-1-benzopyran-4-one
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
35244-11-2
C15H12O3
MFCD00017702
2r-2-2-hydroxyphenyl-2,3-dihydrochromen-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, 2r-2-2-hydroxyphenyl-3,4-dihydro-2h-1-benzopyran-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-1-benzopyran-4-one

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2'-Hydroxyflavanone
Menge 1 g
Formelmasse 240.25
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'