missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylpyridin-4-carbonsäure, 97 %, Thermo Scientific™

Catalog Number p-7030169
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
86454-13-9
C7H7NO3
153.14
MFCD02682019
NQXYVTIQDNOHFM-UHFFFAOYSA-N
2-hydroxy-6-methylisonicotinic acid, 2-hydroxy-6-methylpyridine-4-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxy-6-methyl-isonicotinicacid, 6-hydroxy-2-methylpyridine-4-carboxylic acid, 1,2-dihydro-6-methyl-2-oxopyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 4-pyridinecarboxylicacid, 1,2-dihydro-6-methyl-2-oxo, maybridge3_004231, acmc-209q9q
276660
2-Methyl-6-Oxo-1H-Pyridin-4-Carbonsäure
CC1=CC(=CC(=O)N1)C(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
86454-13-9
C7H7NO3
153.14
MFCD02682019
NQXYVTIQDNOHFM-UHFFFAOYSA-N
2-hydroxy-6-methylisonicotinic acid, 2-hydroxy-6-methylpyridine-4-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxy-6-methyl-isonicotinicacid, 6-hydroxy-2-methylpyridine-4-carboxylic acid, 1,2-dihydro-6-methyl-2-oxopyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 4-pyridinecarboxylicacid, 1,2-dihydro-6-methyl-2-oxo, maybridge3_004231, acmc-209q9q
276660
2-Methyl-6-Oxo-1H-Pyridin-4-Carbonsäure
CC1=CC(=CC(=O)N1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Hydroxy-6-methylpyridine-4-carboxylic acid
Schmelzpunkt >300°C (decomposition)
Menge 1 g
Formelmasse 153.15
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'