missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorbenzonitril, 99 %, Thermo Scientific Chemicals

Catalog Number p-7021862
Click to view available options
Menge:
10 g
50 g
100 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
394-47-8
C7H4FN
121.11
MFCD00001773
GDHXJNRAJRCGMX-UHFFFAOYSA-N
o-fluorobenzonitrile, benzonitrile, 2-fluoro, o-cyanofluorobenzene, 2-fluorobenzonitril, 2-cyanofluorobenzene, benzonitrile, o-fluoro, fluorobenzonitrile, 2-fluorobenzenecarbonitrile, 2-fluoro-benzonitrile, 1-cyano-2-fluorobenzene
67855
2-fluorobenzonitrile
FC1=CC=CC=C1C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
394-47-8
C7H4FN
121.11
MFCD00001773
GDHXJNRAJRCGMX-UHFFFAOYSA-N
o-fluorobenzonitrile, benzonitrile, 2-fluoro, o-cyanofluorobenzene, 2-fluorobenzonitril, 2-cyanofluorobenzene, benzonitrile, o-fluoro, fluorobenzonitrile, 2-fluorobenzenecarbonitrile, 2-fluoro-benzonitrile, 1-cyano-2-fluorobenzene
67855
2-fluorobenzonitrile
FC1=CC=CC=C1C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluorobenzonitrile
Dichte 1.138
Siedepunkt 196°C to 198°C
Flammpunkt 73°C (163°F)
Geruch Almond-like
Brechungsindex 1.507
Menge 10 g
Beilstein 2042184
Formelmasse 121.11
Reinheit (%) 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'