missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoralphamethyl-4-Biphenylessigsäure, 99 %, Thermo Scientific Chemicals

Catalog Number p-7026551
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5104-49-4
C15H13FO2
244.27
MFCD00079303
SYTBZMRGLBWNTM-UHFFFAOYNA-N
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
3394
CHEBI:5130
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5104-49-4
C15H13FO2
244.27
MFCD00079303
SYTBZMRGLBWNTM-UHFFFAOYNA-N
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
3394
CHEBI:5130
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluoro-alpha-methyl-4-biphenylacetic acid
Schmelzpunkt 112°C to 114°C
Geruch Odorless
Menge 1 g
UN-Nummer UN2811
Merck Index 14,4199
IUPAC-Name 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Formelmasse 244.27
Reinheit (%) 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'