missing translation for 'onlineSavingsMsg'
Learn More

2-Fluor-4-(pentafluorthio)benzonitril, 97 %

Catalog Number 15426747
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240256-91-0
C7H3F6NS
247.158
MFCD16652501
OVDJLPWMXSFOJG-UHFFFAOYSA-N
2-fluoro-4-pentafluorosulfur benzonitrile, 2-fluoro-4-pentafluoro-??-sulfanyl benzonitrile, 2-fluoro-4-pentafluorothio benzonitrile
66523489
2-fluor-4-(pentafluor-$l^{6}-sulfanyl)benzonitril
C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240256-91-0
C7H3F6NS
247.158
MFCD16652501
OVDJLPWMXSFOJG-UHFFFAOYSA-N
2-fluoro-4-pentafluorosulfur benzonitrile, 2-fluoro-4-pentafluoro-??-sulfanyl benzonitrile, 2-fluoro-4-pentafluorothio benzonitrile
66523489
2-fluor-4-(pentafluor-$l^{6}-sulfanyl)benzonitril
C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluoro-4-(pentafluorothio)benzonitrile
Menge 1 g
UN-Nummer UN3439
Formelmasse 247.16
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'