missing translation for 'onlineSavingsMsg'
Learn More

2-Fluor-4-Methylbenzenboronsäure, 97 %, Thermo Scientific™

Catalog Number 15470018
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
170981-26-7
C7H8BFO2
153.947
MFCD05664239
LIXXGOMAGHXIMP-UHFFFAOYSA-N
2-fluoro-4-methylphenyl boronic acid, 2-fluoro-4-methylbenzeneboronic acid, 2-fluoro-4-methyl-phenyl boronic acid, boronic acid, 2-fluoro-4-methylphenyl, 2-fluoro-p-tolylboronic acid, 4-methyl-2-fluorophenylboronic acid, boronic acid, b-2-fluoro-4-methylphenyl, pubchem23775, ksc489k8t, 2-fluoro-4methylphenylboronic acid
2783132
(2-Fluor-4-Methylphenyl)boronsäure
B(C1=C(C=C(C=C1)C)F)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
170981-26-7
C7H8BFO2
153.947
MFCD05664239
LIXXGOMAGHXIMP-UHFFFAOYSA-N
2-fluoro-4-methylphenyl boronic acid, 2-fluoro-4-methylbenzeneboronic acid, 2-fluoro-4-methyl-phenyl boronic acid, boronic acid, 2-fluoro-4-methylphenyl, 2-fluoro-p-tolylboronic acid, 4-methyl-2-fluorophenylboronic acid, boronic acid, b-2-fluoro-4-methylphenyl, pubchem23775, ksc489k8t, 2-fluoro-4methylphenylboronic acid
2783132
(2-Fluor-4-Methylphenyl)boronsäure
B(C1=C(C=C(C=C1)C)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluoro-4-methylbenzeneboronic acid
Schmelzpunkt 228°C to 233°C
Menge 1 g
Formelmasse 153.95
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'