missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 2-Cl-IB-MECA
Click to view available options
Menge:
10 mg
50 mg
Unit Size:
10 Milligramm
50 Milligramm
Description
High affinity and extremely selective A3 adenosine receptor agonist (Ki = 0.33 nM). Displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively. Exhibits high selectivity over the Na+-independent adenosine transporter.
Specifications
Specifications
| Chemischer Name oder Material | 2-Cl-IB-MECA |
| Menge | 10 mg |
| Formelmasse | 544.74 |
| Reinheit | >98% |
| Molekulargewicht | 544.734 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction