missing translation for 'onlineSavingsMsg'
Learn More

2-Chlormethyl-3,4-dimethoxypyridinhydrochlorid, 97 %, Thermo Scientific Chemicals

Artikelnummer. 15415586
Change view
Click to view available options
Menge:
1 g
5 g
Unit Size:
1 Gramm
5 Gramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
15415586 5 g 5 Gramm
15405586 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415586 Supplier Thermo Scientific Alfa Aesar Supplier No. H60235.06
Bulk oder benutzerdefiniertes Format anfordern

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Dieses Markenprodukt von Thermo Scientific war ursprünglich Teil des Produktportfolios von Alfa Aesar.Einige Dokumente und Etiketteninformationen nennen möglicherweise die bisherige Marke.Die ursprüngliche Produkt- oder Artikelnummer bei Alfa Aesar oder die SKU-Referenz wurde im Rahmen des Übergangs der Marke zu Thermo Scientific nicht geändert.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72830-09-2
Summenformel C8H11Cl2NO2
Molekulargewicht (g/mol) 224.081
MDL-Nummer MFCD02181083
InChI-Schlüssel YYRIKJFWBIEEDH-UHFFFAOYSA-N
Synonym 2-chloromethyl-3,4-dimethoxypyridine hydrochloride, 2-chloromethyl-3,4-dimethoxy pyridine hydrochloride, 2-chloromethyl-3,4-dimethoxypyridinium chloride, 2-chloromethyl-3,4-dimethoxypyridinium hydrochloride, 2-chloromethyl-3 pound not4-dimethoxy-pyridine hydrochloride, 2-chloromethyl-3,4-dimethoxy pyridine hcl, 3,4-dimethoxy-2-chloromethylpyridine hydrochloride, 2-chloromethyl-3,4-dimethoxypyridine;hydrochloride, 2-chloromethyl-3,4-dimethoxy-pyridine hydrochloride
PubChem CID 16216928
IUPAC-Name 2-Chlormethyl-3,4-Dimethoxypyridin;hydrochlorid
SMILES COC1=C(C(=NC=C1)CCl)OC.Cl

Specifications

Chemischer Name oder Material 2-Chloromethyl-3,4-dimethoxypyridine hydrochloride
Schmelzpunkt 155°C (decomposition)
Menge 5 g
UN-Nummer UN3077
Formelmasse 224.08
Reinheit (%) 97%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.