missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-4-Fluorbenzenboronsäure, 98 %, Thermo Scientific Chemicals

Catalog Number p-7030059
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
313545-72-1
C6H5BClFO2
174.36
MFCD02684295
XOFNMNLYGPKKOV-UHFFFAOYSA-N
2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
17750229
(2-chloro-4-fluorophenyl)boronic acid
OB(O)C1=CC=C(F)C=C1Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
313545-72-1
C6H5BClFO2
174.36
MFCD02684295
XOFNMNLYGPKKOV-UHFFFAOYSA-N
2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
17750229
(2-chloro-4-fluorophenyl)boronic acid
OB(O)C1=CC=C(F)C=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-4-fluorobenzeneboronic acid
Menge 1 g
Formelmasse 174.37
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'