Learn More
2-Chlor-3-cyanpyridin-4-boronsäurepinakolester, 95 %, Thermo Scientific™
Catalog Number 15437675
Click to view available options
Menge:
250 mg
1 g
878194-94-6
C12H14BClN2O2
264.516
MFCD09037479
YYPZHORJYDCOSQ-UHFFFAOYSA-N
2-chloro-3-cyanopyridine-4-boronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 2-chloro-3-cyano-4-pyridinylboronoic acid pinacol, 2-chloro-3-cyano-4-pyridinylboronic acid pinacol ester, 2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile
44755174
2-Chlor-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Pyridin-3-Carbonitril
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N
878194-94-6
C12H14BClN2O2
264.516
MFCD09037479
YYPZHORJYDCOSQ-UHFFFAOYSA-N
2-chloro-3-cyanopyridine-4-boronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 2-chloro-3-cyano-4-pyridinylboronoic acid pinacol, 2-chloro-3-cyano-4-pyridinylboronic acid pinacol ester, 2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile
44755174
2-Chlor-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Pyridin-3-Carbonitril
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester
Schmelzpunkt 100°C to 101°C
Menge 250 mg
Formelmasse 264.52
Reinheit (%) 95%