missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-4-(2-furyl)-thiazol, 97 %, Thermo Scientific™

Catalog Number 15454638
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
917566-86-0
C7H4BrNOS
230.079
MFCD11220134
PUFWLSRDFTVPMS-UHFFFAOYSA-N
2-bromo-4-furan-2-yl-1,3-thiazole, 2-bromo-4-2-furyl thiazole, 2-bromo-4-2-furanyl thiazole, 2-bromo-4-furan-2-yl thiazole, thiazole, 2-bromo-4-2-furanyl
38996209
2-Brom-4-(Furan-2-yl)-1,3-Thiazol
C1=COC(=C1)C2=CSC(=N2)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
917566-86-0
C7H4BrNOS
230.079
MFCD11220134
PUFWLSRDFTVPMS-UHFFFAOYSA-N
2-bromo-4-furan-2-yl-1,3-thiazole, 2-bromo-4-2-furyl thiazole, 2-bromo-4-2-furanyl thiazole, 2-bromo-4-furan-2-yl thiazole, thiazole, 2-bromo-4-2-furanyl
38996209
2-Brom-4-(Furan-2-yl)-1,3-Thiazol
C1=COC(=C1)C2=CSC(=N2)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-4-(2-furyl)thiazole
Menge 1 g
Beilstein 15461854
Formelmasse 230.08
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'