missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibrombenzoesäure, 95 %, Thermo Scientific Chemicals

Catalog Number p-7077721
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
601-84-3
C7H4Br2O2
279.92
MFCD00234254
HQLOEBRPCVIFCT-UHFFFAOYSA-N
benzoic acid, 2,6-dibromo, 2,6-dibromobenzoicacid, 2,6-bis bromanyl benzoic acid
4400143
2,6-dibromobenzoic acid
OC(=O)C1=C(Br)C=CC=C1Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
601-84-3
C7H4Br2O2
279.92
MFCD00234254
HQLOEBRPCVIFCT-UHFFFAOYSA-N
benzoic acid, 2,6-dibromo, 2,6-dibromobenzoicacid, 2,6-bis bromanyl benzoic acid
4400143
2,6-dibromobenzoic acid
OC(=O)C1=C(Br)C=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,6-Dibromobenzoic acid
Schmelzpunkt 141.0°C to 142.0°C
Infrarotspektrum Authentic
Prozentgehaltsbereich 94% min. (HPLC)
Verpackung Glasflasche
Menge 1 g
Formelmasse 279.92
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'