missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-3-thiophensulfonylchlorid, 97 %, Thermo Scientific™
Click to view available options
Menge:
1 g
5 g
10 g
Spezifikation
Chemischer Name oder Material | 2, 5-Dimethyl-3-thiophenesulfonyl chloride |
CAS Min. % | 97.0 |
Prozentgehaltsbereich | 97% |
Verpackung | Bernsteinfarbene Glasflasche |
Menge | 1 g |
Synonym | 2,5-dimethyl-3-thiophenesulfonyl chloride, 2,5-dimethylthiophene-3-sulphonyl chloride, 3-thiophenesulfonylchloride, 2,5-dimethyl, buttpark 9650-20, chembrdg-bb 4009594, 2,5-dimethyl 3-thienyl chlorosulfone, 2,5-dimethyl-3-thienyl sulfonyl chloride, 2, 5-dimethylthiophene-3-sulphonyl chloride, 3-thiophenesulfonylchloride,2,5-dimethyl-9ci |
Formelmasse | 210.7 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'