missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)-imidazol-4,5-dithiocarboxamid, 97 %, Thermo Scientific™

Catalog Number p-7036578
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1347815-08-0
C12H12N4S2
276.376
MFCD20265364
CFNPTMBECAYGCK-UHFFFAOYSA-N
2-4-methylphenyl imidazole-4,5-dithiocarboxamide, 2-4-methylphenyl-1h-imidazole-4,5-dicarbothioamide, 2-p-tolyl imidazole-4,5-dithiocarboxamide
73996015
2-(4-Methylphenyl)-1H-Imidazol-4,5-Dicarbothioamid
CC1=CC=C(C=C1)C2=NC(=C(N2)C(=S)N)C(=S)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1347815-08-0
C12H12N4S2
276.376
MFCD20265364
CFNPTMBECAYGCK-UHFFFAOYSA-N
2-4-methylphenyl imidazole-4,5-dithiocarboxamide, 2-4-methylphenyl-1h-imidazole-4,5-dicarbothioamide, 2-p-tolyl imidazole-4,5-dithiocarboxamide
73996015
2-(4-Methylphenyl)-1H-Imidazol-4,5-Dicarbothioamid
CC1=CC=C(C=C1)C2=NC(=C(N2)C(=S)N)C(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(4-Methylphenyl)imidazole-4,5-dithiocarboxamide
Menge 250 mg
Formelmasse 276.37
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'