missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorbenzylalkohol, 97 %, Thermo Scientific™

Catalog Number p-7025361
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
118289-07-9
C7H5F3O
162.111
MFCD00061207
YJQDVBPZXNALEE-UHFFFAOYSA-N
2,4,6-trifluorobenzyl alcohol, 2,4,6-trifluorophenyl methanol, 2,4,6-trifluorobenzylalcohol, benzenemethanol,2,4,6-trifluoro, 2,4,6-trifluorophenyl methan-1-ol, acmc-20aof2, 2,4,6-trifluoro-phenyl-methanol, 2,4,6-trifluorophenylmethyl alcohol, 2,4,6-tris fluoranyl phenyl methanol
2777037
(2,4,6-trifluorphenyl)methanol
C1=C(C=C(C(=C1F)CO)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
118289-07-9
C7H5F3O
162.111
MFCD00061207
YJQDVBPZXNALEE-UHFFFAOYSA-N
2,4,6-trifluorobenzyl alcohol, 2,4,6-trifluorophenyl methanol, 2,4,6-trifluorobenzylalcohol, benzenemethanol,2,4,6-trifluoro, 2,4,6-trifluorophenyl methan-1-ol, acmc-20aof2, 2,4,6-trifluoro-phenyl-methanol, 2,4,6-trifluorophenylmethyl alcohol, 2,4,6-tris fluoranyl phenyl methanol
2777037
(2,4,6-trifluorphenyl)methanol
C1=C(C=C(C(=C1F)CO)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,4,6-Trifluorobenzyl alcohol
Menge 1 g
Formelmasse 162.11
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'