missing translation for 'onlineSavingsMsg'
Learn More

2',3 ',6 '-Trifluorpropiophenon, 97+ %

Catalog Number 11445887
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
243666-18-4
C9H7F3O
188.15
MFCD00061222
LEDVCRMPYBDECF-UHFFFAOYSA-N
2',3',6'-trifluoropropiophenone, 1-2,3,6-trifluorophenyl propan-1-one, 2,3,6-trifluoropropiophenone, acmc-1cg4s, 1-propanone,1-2,3,6-trifluorophenyl
2777975
1-(2,3,6-trifluorophenyl)propan-1-one
CCC(=O)C1=C(F)C=CC(F)=C1F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
243666-18-4
C9H7F3O
188.15
MFCD00061222
LEDVCRMPYBDECF-UHFFFAOYSA-N
2',3',6'-trifluoropropiophenone, 1-2,3,6-trifluorophenyl propan-1-one, 2,3,6-trifluoropropiophenone, acmc-1cg4s, 1-propanone,1-2,3,6-trifluorophenyl
2777975
1-(2,3,6-trifluorophenyl)propan-1-one
CCC(=O)C1=C(F)C=CC(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2',3',6'-Trifluoropropiophenone
Menge 1 g
Formelmasse 188.15
Reinheit (%) ≥97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'