Learn More
Tocris Bioscience™ 1S,3R-RSL3
Click to view available options
Menge:
10 mg
50 mg
Packungsgröße:
10 Milligramm
50 Milligramm
Beschreibung
1S,3R-RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.
Negative control 1R,3R-RSL3 (Cat. No. 6687) also available.
Spezifikation
Spezifikation
| Chemischer Name oder Material | (1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate |
| CAS | 1219810-16-8 |
| Prozentgehaltsbereich | >98% |
| Menge | 10 mg |
| Summenformel | C23H21ClN2O5 |
| Formelmasse | Observed MW: 440.88 |