Learn More
Thermo Scientific Alfa Aesar 1KG Diaion|r WA10, ion exchange resin, weakly basic gel type, 1.2 meq/ml on polyacrylate 1kg
Marke: Thermo Scientific Alfa Aesar 046700.A1
1KG Diaion|r WA10, ion exchange resin, weakly basic gel type, 1.2 meq/ml on polyacrylate 1kg
Additional Details : CAS : 63182-01-4 Gewicht : 1.00000kg