Learn More
Agilent BioTek 1536-well Microplate Wash Aspiration Manifold Kit
Microplate Wash Aspiration Manifold Kit for use with the 406 FX Washer Dispenser.
Marke: Agilent BioTek 2120085
Additional Details : Gewicht : 4.08233kg
Beschreibung
- Includes aspiration manifold, alignment jig and instructions
- Aspirate-only manifold is used with either a dual syringe pump dispenser module and 32-tube dispense manifold(s), or with a peristaltic pump dispenser module and a 1536-well microplate cassette
- Dispense manifolds and cassettes are ordered separately
- For syringe pump dispensing into 1536-well plates, at least one 32-tube dispense manifold is required
- For faster dispensing, two 32-tube dispense manifolds can be used simultaneously
Spezifikation
Wash Module | |
406 FX Washer Dispenser |