missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11648-2-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838873

  • 131.19 CHF / 20 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody was raised against full-length 14-3-3 epsilon.

This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the mouse ortholog. It interacts with CDC25 phosphatases, RAF1 and IRS1 proteins, suggesting its role in diverse biochemical activities related to signal transduction, such as cell division and regulation of insulin sensitivity. It has also been implicated in the pathogenesis of small cell lung cancer. Two transcript variants, one protein-coding and the other non-protein-coding, have been found for this gene.
Spezifikation

Spezifikation

14-3-3 epsilon
Polyclonal
Unconjugated
YWHAE
14 3 3 protein epsilon, 14 3 3E, 14-3-3, 14-3-3 epsilon, KCIP 1, MDCR, MDS, YWHAE
Rabbit
Antigen Affinity Chromatography
RUO
22627, 29753, 7531
Antibody
IgG
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunocytochemistry
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P62258, P62259, P62260
YWHAE
14-3-3 epsilon Fusion Protein Ag2247
20 μL
Primary
Human, Rat, Mouse
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote